Learn More
Cole-Parmer Cleanroom Female Luer to Hose Barb Fittings, Straight Adapter, Natural Nylon
Product Code. 30053072 Shop All Cole-Parmer Products
Click to view available options
Outlet Port:
1/16 in. Hose Barb
1/4 in. Hose Barb
1/8 in. Hose Barb
3/16 in. Hose Barb
3/32 in. Hose Barb
5/32 in. Hose Barb
Unit Size:
Pack of 10
Product Code. 30053072
Brand: Cole-Parmer 5010994
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn. Single barb design tightly grips the tubing. RoHS compliant
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Material Nylon
Packaging Quantity 10/Pk.
Inlet Female Luer x Hose Barb
Outlet Port 1/4 in. Hose Barb
Color Natural