Learn More
Cole-Parmer Cleanroom Female Luer to Hose Barb Fittings, Straight Adapter, Natural Kynar™
Shop All Cole-Parmer Products
Click to view available options
Outlet Port:
1/16 in. Hose barb
1/4 in. Hose barb
1/8 in. Hose barb
5/32 in. Hose barb
Unit Size:
Pack of 10
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;USP |
| Material | Kynar |
| Packaging Quantity | 10/Pk. |
| Inlet | Female Luer x Hose Barb |
| Outlet Port | 1/8 in. Hose barb |
| Color | Natural |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.