Learn More
Cole-Parmer Cleanroom Female Luer Fittings, Straight Union
Product Code. 30054368 Shop All Cole-Parmer Products
Change view
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nylon
Polypropylene
Color:
Clear
Natural
White
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Color unitSize
30054368 Nylon White Pack of 10
30053726 Polypropylene Clear Pack of 10
30054155 Kynar Natural Pack of 10
30054282 Nylon Natural Pack of 10
30054204 CrystalVu (CVU) Clear Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30054368 Supplier Cole-Parmer Supplier No. 5011422
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Forms a leak-free luer connection with a quick simple turn. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Certifications/Compliance FDA;Non-DEHP;Non-Phthalate;RoHS
Material Nylon
Packaging Quantity 10/Pk.
Inlet Female Luer
Color White