missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Branson Ultrasonics™ Cleaning Reagent, General Purpose
Description
- Fingerprints
- Dust
- Packaging particulates
- Light oils and greases from metal working and light machine operations.
- Metal stampings and machined components
- Glassware
- Rigid injection molded parts
- Other components with light-to-intermediate soil levels
Specifications
Specifications
| Product Type | Cleaning Reagent |
| pH | 13 |
| For Use With (Application) | For General-purpose use |
| Volume (Metric) | 1 L |
Safety and Handling
Product Identifier
- General Purpose
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 2
- H314-Causes severe skin burns and eye damage.
- H319-Causes serious eye irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: natriummetasilicat pentahydrat, 2-butoxyethanol, isotridecanol ethoxylated, sodium 2-(2-dodecyloxyet
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?