missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Cleaning Pen
Click to view available options
Color:
White
Unit Size:
Each
Description
- For use with Global Mark, Powermark Printers.
Specifications
Specifications
| Product Type | Cleaning Pen |
| Color | White |
| For Use With (Equipment) | GlobalMark Label Maker, PowerMark Industrial Sign and Label Printer |
| Description | Cleaning Pen for DIY systems |
| Format | Cleaner |
| Quantity | 1 Ea. |
| Alternate Part Number | Cleaning Pen |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction