missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Cleaning Detergent rea-phos™

Product Code. 10565402
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10565402 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10565402 Supplier Hirschmann™ Supplier No. 9740101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For rapid, automatic and careful cleaning of materials such as glass, porcelain, rubber, plastics and metals. Hirschmann™ rea-phos™ Phosphate-free Rapid Cleaning Agent is ideal for the laboratory in removing oil, grease, wax, resin, silicone, blood, protein and serum residues.

  • Phosphate-free
  • Removes oil, fat, blood and protein residues
  • Usable in hot water
  • Suitable for ultrasonic systems

Specifications

Product Type Phosphate-free Rapid Cleaning Concentrate
Form Liquid
Product Line rea-phos
Concentration 1%
Description Biodegradable
Quantity 1 Bottle
Volume (Metric) 1 L
compliance-icons
Product Identifier
  • rea-phos
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P390-Absorb spillage to prevent material damage.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.