missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Cleaning Brush Set
Description
- Cleaning Brushes for for the BSP™31 Labels Attachment System.
Specifications
Specifications
| Product Type | Brush Kit |
| Color | Black |
| For Use With (Equipment) | BSP31 Printer |
| Description | Cleaning Brushes for the BSP31 |
| Format | Brush kit |
| For Use With (Application) | Laboratory labelling |
| Quantity | 1 Ea. |
| Alternate Part Number | BSP31-BRUSH |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?