Learn More
Phenomenex™ Cleanert™ Screw Caps with Septa, 9 mm, Pre-Slit
Product Code. 31748037 Shop All Phenomenex Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 100
Product Code. Color Unit Size
31748037 Blue Pack of 100
1 options
Product Code. 31748037 Supplier Phenomenex™ Supplier No. AV22020
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accessories for use with chromatography vials and in various chromatography applications.
  • Brand: Cleanert
  • Product: Accessory
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Diameter (Metric) Outer 9 mm
Pre-slit No
Sterility Sterile
Septum PTFE/Silicone
Closure Size 9 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap