missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLE7 homolog Antibody (OTI2A4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71940CL1
This item is not returnable.
View return policy
Description
CLE7 homolog Monoclonal antibody specifically detects CLE7 homolog in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| CLE7 homolog | |
| Monoclonal | |
| CoraFluor 1 | |
| CGI99, CGI-99, CLE, LCRP369, RLLM1 | |
| Full length human recombinant protein of human C14orf166 (NP_057123) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 51637 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI2A4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction