missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLCNKB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119867
This item is not returnable.
View return policy
Description
Positive Control: Mouse testis tissue lysate, rat testis tissue lysate, MCF-7 cell lysate, human kidney tissue, human seminoma tissue, MG-63, A549. Subcellular Location: Cell membrane.
Chloride channel Kb (CLCNKB) is a member of the CLC family of voltage-gated chloride channels, which comprises at least 9 mammalian chloride channels. Each is believed to have 12 transmembrane domains and intracellular N and C termini. Mutations in CLCNKB result in the autosomal recessive Type III Bartter Syndrome. CLCNKA and CLCNKB are closely related (94% sequence identity), tightly linked (separated by 11 kb of genomic sequence) and are both expressed in mammalian kidney.
Specifications
| CLCNKB | |
| Polyclonal | |
| Unconjugated | |
| CLCNKB | |
| chloride channel; chloride channel K1-like; putative basolateral cTAL chloride channel ClC-Ka; chloride channel K2; chloride channel Ka; Chloride channel Kb; chloride channel protein ClC-Ka; Chloride channel protein ClC-Kb; chloride channel, kidney, B; chloride channel, voltage-sensitive Ka; chloride channel, voltage-sensitive Kb; chloride voltage-gated channel Kb; clC-K1; Clck2; ClC-K2; ClC-K2L; Clc-Ka; CLCKB; ClC-Kb; CLCNK; Clcnk1l; CLCNKA; Clcnkb; hClC-Kb; LOC100730738; MGC24087; putative basolateral mTAL chloride channel ClC-Ka; RP11-5P18.8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1188, 56365, 79430 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P51801, P51802, Q9WUB6 | |
| CLCNKB | |
| Recombinant protein within C-terminal human CLCNKB from aa520-aa687. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction