missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCNKA Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £359.00
Specifications
| Antigen | CLCNKA |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18656422
|
Novus Biologicals
NBP2-92433-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699161
|
Novus Biologicals
NBP2-92433-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLCNKA Polyclonal antibody specifically detects CLCNKA in Human, Mouse samples. It is validated for Western BlotSpecifications
| CLCNKA | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 1187 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| chloride channel KaMGC61490, hClC-Ka, kidney, A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 470-644 of human CLCNKA (NP_001244068.1). QSCQPSFYDGTIIVKKLPYLPRILGRNIGSHHVRVEHFMNHSITTLAKDTPLEEVVKVVTSTDVTEYPLVESTESQILVGIVQRAQLVQALQAEPPSRAPGHQQCLQDILARGCPTEPVTLTLFSETTLHQAQNLFKLLNLQSLFVTSRGRAVGCVSWVEMKKAISNLTNPPAPK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title