missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA560426
This item is not returnable.
View return policy
Description
Immunogen sequence: LRLKDFRDAY PRFPPIQSIH VSQDERECTM DLSEFMNPSP YTVPQEASLP RVFKLFRALG LRHLVVVDNR NQVVGLVT Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
The product of this gene belongs to the CLC chloride channel family of proteins. Chloride channels play important roles in the plasma membrane and in intracellular organelles. This gene encodes chloride channel 7. Defects in this gene are the cause of osteopetrosis autosomal recessive type 4 (OPTB4), also called infantile malignant osteopetrosis type 2 as well as the cause of autosomal dominant osteopetrosis type 2 (OPTA2), also called autosomal dominant Albers-Schonberg disease or marble disease autosoml dominant. Osteopetrosis is a rare genetic disease characterized by abnormally dense bone, due to defective resorption of immature bone. OPTA2 is the most common form of osteopetrosis, occurring in adolescence or adulthood.
Specifications
| CLCN7 | |
| Polyclonal | |
| Unconjugated | |
| CLCN7 | |
| AA409691; AW538136; chloride channel 7; chloride channel 7 alpha subunit; Chloride channel protein 7; chloride channel, voltage-sensitive 7; chloride voltage-gated channel 7; CLC7; CLC-7; CLCN7; CLCN-7; D17Wsu51e; H(+)/Cl(-) exchange transporter 7; OPTA2; OPTB4; PPP1R63; protein phosphatase 1, regulatory subunit 63 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1186 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51798 | |
| CLCN7 | |
| Recombinant protein corresponding to Human CLCN7 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction