missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92800-0.1ml
This item is not returnable.
View return policy
Description
CLCN7 Polyclonal antibody specifically detects CLCN7 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CLCN7 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| chloride channel 7, Chloride channel protein 7, CLC-7, CLC7FLJ39644, FLJ26686, H(+)/Cl(-) exchange transporter 7, OPTA2FLJ46423, OPTB4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 626-805 of human CLCN7 (NP_001278.1). TAREVMSTPVTCLRRREKVGVIVDVLSDTASNHNGFPVVEHADDTQPARLQGLILRSQLIVLLKHKVFVERSNLGLVQRRLRLKDFRDAYPRFPPIQSIHVSQDERECTMDLSEFMNPSPYTVPQEASLPRVFKLFRALGLRHLVVVDNRNQVVGLVTRKDLARYRLGKRGLEELSLAQT | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 1186 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction