missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539101
This item is not returnable.
View return policy
Description
This gene encodes a member of the voltage-dependent chloride channel protein family. Members of this family can function as either chloride channels or antiporters. This protein is primarily localized to late endosomes and functions as a chloride/proton antiporter. Alternate splicing results in both coding and non-coding variants. Additional alternately spliced variants have been described but their full-length structure is unknown. [provided by RefSeq, Mar 2012].
Specifications
| CLCN6 | |
| Polyclonal | |
| Unconjugated | |
| CLCN6 | |
| D, ClC-6d, D1-A2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1185, 26372 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51797, O35454 | |
| CLCN6 | |
| A synthetic peptide derived from the internal region of human CLCN6 | |
| 100 μg | |
| Primary | |
| Human, Mouse, Non-human Primate | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction