missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLCN4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550933
This item is not returnable.
View return policy
Description
CLC-4 Polyclonal Antibody detects endogenous levels of CLC-4 protein.
The family of voltage-dependent chloride channels (CLCs) regulate cellular trafficking of chloride ions, a critical component of all living cells. CLCs regulate excitability in muscle and nerve cells, aid in organic solute transport and maintain cellular volume. The genes encoding human CLC-1 through CLC-7 map to chromosomes 7, 3q26, 4q32, Xp22, Xp11, 1p36 and 16p13, respectively. CLC-1 is highly expressed in skeletal muscle. Mutations in the gene encoding CLC-1 lead to myotonia, an inheritable disorder characterized by muscle stiffness and renal salt wasting. CLC-2 is highly expressed in the epithelia of several organs including lung, which suggests CLC-2 may be a possible therapeutic target for cystic fibrosis. CLC-3 expression is particularly abundant in neuronal tissue, while CLC-4 expression is evident in skeletal and cardiac muscle as well as brain.
Specifications
| CLCN4 | |
| Polyclonal | |
| Unconjugated | |
| CLCN4 | |
| chloride channel 4; chloride channel 4-2; chloride channel protein 4; chloride channel, voltage-sensitive 4; chloride transporter ClC-4; chloride voltage-gated channel 4; CLC 4; CLC4; clC-4; Clc4-2; ClC-4A; CLCN4; Clcn4-2; H(+)/Cl(-) exchange transporter 4; MGC163150; putative chloride channel (similar to Mm Clcn4-2); putative chloride channel 4-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1183, 12727, 60586 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| P51793, P51794, Q61418 | |
| CLCN4 | |
| Synthesized peptide derived from the Internal region of human CLC-4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction