missing translation for 'onlineSavingsMsg'
Learn More

CLCN3 Antibody (S258-5), Novus Biologicals™

Product Code. 18202492 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18202492 100 μg 100µg
18691037 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18202492 Supplier Novus Biologicals Supplier No. NBP259379

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CLCN3 Monoclonal specifically detects CLCN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCN3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S258-5
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
Gene Alias chloride channel 3, Chloride channel protein 3, Chloride transporter ClC-3, ClC-3, CLC3, H(+)/Cl(-) exchange transporter 3
Gene Symbols CLCN3
Host Species Mouse
Immunogen Synthetic peptide amino acids 98-115 (cytoplasmic N-terminus) of rat Clcn3
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1182
Test Specificity Detects 90kDa. Does not cross-react with Clcn4 or Clcn5 (based on KO validation results).
Target Species Human, Mouse, Rat
Content And Storage Store at -20C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.