missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCN2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-52032
This item is not returnable.
View return policy
Description
CLCN2 Polyclonal specifically detects CLCN2 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| CLCN2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| chloride channel 2, chloride channel protein 2, CIC-2, clC-2, CLC2, ECA2, ECA3, EGI11, EGI3, EGMA, EJM6, EJM8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1181 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| NP_004357.3 | |
| CLCN2 | |
| Peptide with sequence QQLDEPVNFSDCK corresponding to internal region according to NP_004357.3. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize all reported isoforms (NP_004357.3; NP_001164558.1; NP_001164559.1; NP_001164560.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction