missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCA4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92365-0.1ml
This item is not returnable.
View return policy
Description
CLCA4 Polyclonal antibody specifically detects CLCA4 in Human, Rat samples. It is validated for Western Blot
Specifications
| CLCA4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| caCC-2, CaCC2CaCC, Calcium-activated chloride channel family member 4, Calcium-activated chloride channel protein 2, calcium-activated chloride channel regulator 4, chloride channel accessory 4, chloride channel regulator 4, chloride channel, calcium activated, family member 4, hCaCC-2, hCLCA4, MGC142247, MGC142249 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 22-150 of human CLCA4 (NP_036260.2). SFIKLNNNGFEDIVIVIDPSVPEDEKIIEQIEDMVTTASTYLFEATEKRFFFKNVSILIPENWKENPQYKRPKHENHKHADVIVAPPTLPGRDEPYTKQFTECGEKGEYIHFTPDLLLGKKQNEYGPPG | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 22802 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction