missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLCA3P Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549089
This item is not returnable.
View return policy
Description
CLCA3P Polyclonal Antibody for Western Blot
This gene is a transcribed pseudogene belonging to the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same site on chromosome 1p31-p22 and share high degrees of homology in size, sequence and predicted structure, but differ significantly in their tissue distributions. This gene contains several nonsense codons compared to other family members that render the transcript a candidate for nonsense-mediated mRNA decay (NMD). Therefore, this gene is unlikely to be protein-coding. [provided by RefSeq, Jan 2009].
Specifications
| CLCA3P | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9Y6N3 | |
| CLCA3P | |
| KLH conjugated synthetic peptide between 70-98 amino acids from the Central region of human CLCA3P. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| CLCA3P | |
| chloride channel accessory 3, pseudogene; CLCA3; CLCA3P; hCLCA3 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 9629 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction