missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCA2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19273-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the calcium sensitive chloride conductance protein family. To date, all members of this gene family map to the same site on chromosome 1p31-p22 and share high degrees of homology in size, sequence and predicted structure, but differ significantly in their tissue distributions. Since this protein is expressed predomitly in trachea and lung, it is suggested to play a role in the complex pathogenesis of cystic fibrosis. It may also serve as adhesion molecule for lung metastatic cancer cells, mediating vascular arrest and colonization, and furthermore, it has been implicated to act as a tumor suppressor gene for breast cancer.Specifications
| CLCA2 | |
| Polyclonal | |
| Unconjugated | |
| CLCA2 | |
| CACC, CaCC 3, CACC3, chloride channel accessory 2, CLCA2, CLCRG2, hCaCC 3, hCLCA2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 229933, 9635 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BG22, Q9UQC9 | |
| CLCA2 | |
| CLCA2 Fusion Protein Ag5914 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction