missing translation for 'onlineSavingsMsg'
Learn More

CLCA2 Antibody (1B9), Novus Biologicals™

Product Code. 18386658 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386658 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386658 Supplier Novus Biologicals Supplier No. H00009635M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CLCA2 Monoclonal antibody specifically detects CLCA2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLCA2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1B9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006527
Gene Alias CACC, CaCC-3, CACC3, Calcium-activated chloride channel family member 2, Calcium-activated chloride channel protein 3, calcium-activated chloride channel regulator 2, calcium-activated chloride channel-2, chloride channel accessory 2, chloride channel regulator 2, chloride channel, calcium activated, family member 2, CLCA family member 2, chloride channel regulator, CLCRG2, FLJ97885, hCaCC-3, hCLCA2
Host Species Mouse
Immunogen CLCA2 (NP_006527, 300 a.a. ~ 400 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PTFSLVQAGDKVVCLVLDVSSKMAEADRLLQLQQAAEFYLMQIVEIHTFVGIASFDSKGEIRAQLHQINSNDDRKLLVSYLPTTVSAKTDISICSGLKKGF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Breast Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9635
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.