missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99573-50ul
This item is not returnable.
View return policy
Description
CLCA1 Polyclonal antibody specifically detects CLCA1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CLCA1 | |
| Polyclonal | |
| Immunohistochemistry 1:1000-1:5000, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| CaCC, Calcium-activated chloride channel family member 1, Calcium-activated chloride channel protein 1, chloride channel accessory 1, chloride channel regulator, chloride channel regulator 1, chloride channel, calcium activated, family member 1, CLCRG1, FLJ95147, GOB5, hCaCC-1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CLCA1. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 1179 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction