missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CLC-5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596402
This item is not returnable.
View return policy
Description
Positive Samples: SW480, U-937, HepG2, A673, NCI-H460, Mouse kidney, Mouse ovary, Mouse brain; Cellular Location: Cell membrane, Endosome membrane, Golgi apparatus membrane, Multi-pass membrane protein.
Proton-coupled chloride transporter. Functions as antiport system and exchanges chloride ions against protons. Important for normal acidification of the endosome lumen. May play an important role in renal tubular function.
Specifications
| CLC-5 | |
| Polyclonal | |
| Unconjugated | |
| CLCN5 | |
| 5430408K11Rik; chloride channel 5; chloride channel 5 (nephrolithiasis 2, X-linked, Dent disease); chloride channel ClC5; chloride channel Clc-5; chloride channel CLCN5; chloride channel protein 5; chloride channel, voltage-sensitive 5; chloride transporter ClC-5; chloride voltage-gated channel 5; Clc5; clC-5; CLCK2; CLCN3; CLCN4; Clcn5; D930009B12Rik; DENTS; DXImx42e; H(+)/Cl(-) exchange transporter 5; H(+)/Cl(-) exchange transporter 5-like; hCIC-K2; hClC-K2; NPHL1; NPHL2; Sfc13; T25545; voltage-gated chloride ion channel CLCN5; XLRH; XRN | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1184, 12728 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51795, Q9WVD4 | |
| CLCN5 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction