Learn More
Emma Safety Footwear Clay (Ruffneck) Safety Shoes
Product Code. 15999071
Change view
Click to view available options
Size:
39
40
41
42
43
45
46
47
48
49
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15999071 39 1 pair
15909081 40 1 pair
15919081 41 1 pair
15929081 42 1 pair
15939081 43 1 pair
15949081 45 1 pair
15959081 46 1 pair
15969081 47 1 pair
15979081 48 1 pair
15989081 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15999071 Supplier Emma Safety Footwear Supplier No. 91654639
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reinforced ruff-neck heel together with the shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • S3-Safety shoe with sole plate for wet work
  • Nubuck / Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • ESD certified
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Clay (Ruffneck)
Liner Material Hydro-Tec™ Sanitized Silver
Material Nubuck, Oiled Nubuck
Size 39
Toe Material Steel