missing translation for 'onlineSavingsMsg'
Learn More

Clavulanic Acid Potassium Salt, TRC

Product Code. 17952205
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17952205 100 mg 100mg
17924015 10 mg 10mg
17940625 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17952205 Supplier Toronto Research Chemicals Supplier No. TRCC563750100MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61177-45-5
Molecular Formula C8 H8 N O5 . K
Molecular Weight (g/mol) 237.25
Synonym 4-Oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3-(2-hydroxyethylidene)-7-oxo-, monopotassium salt, (2R,3Z,5R)- (9CI), 4-Oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3-(2-hydroxyethylidene)-7-oxo-, monopotassium salt, [2R-(2α,3Z,5α)]-, BRL 14151K, Clavitesse, Clavulanate potassium, Clavulanic acid potassium salt, Potassium clavulanate, Serdaxin
IUPAC Name potassium;(2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [K+].OC\C=C\1/O[C@@H]2CC(=O)N2[C@H]1C(=O)[O-]

Specifications

Analyte or Component Names Clavulanic acid potassium salt
Shipping Condition Blue Ice
InChI Formula InChI=1S/C8H9NO5.K/c10-2-1-4-7(8(12)13)9-5(11)3-6(9)14-4;/h1,6-7,10H,2-3H2,(H,12,13);/q;+1/p-1/b4-1-;/t6-,7-;/m1./s1
Solution Type Neat
Industry Type Forensics
Quantity 100 mg
Formula Weight 237.004
Percent Purity >95%
Chemical Name or Material Clavulanic acid potassium salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.