missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-8 Antibody (474210) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5275JF635
This item is not returnable.
View return policy
Description
Claudin-8 Monoclonal antibody specifically detects Claudin-8 in Human samples. It is validated for Flow Cytometry,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Claudin-8 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| claudin 8, human CLDN8 gene for claudin-810claudin-8 | |
| HEK293 human embryonic kidney cell line transfected with human Claudin-8, Met1-Val225 (Ser151Pro), Accession # P56748 | |
| RUO | |
| 9073 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 474210 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction