missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 7 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29795-1-AP-20UL
This item is not returnable.
View return policy
Description
The Claudin 7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Claudin 7. This antibody recognizes human antigen. The Claudin 7 antibody has been validated for the following applications: IHC, ELISA analysis.Specifications
| Claudin 7 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1366 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Immunohistochemistry, ELISA | |
| 900 μg/mL | |
| CEPTRL2; CPETRL2; Hs.84359; claudin-1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 23 kDa | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu