missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 7 Monoclonal Antibody (6-G3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544622
This item is not returnable.
View return policy
Description
Claudin 7 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF
This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions. Differential expression of this gene has been observed in different types of malignancies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.
Specifications
| Claudin 7 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 40% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4 | |
| O95471, Q9Z1L1, Q9Z261 | |
| CLDN7 | |
| Synthetic peptide within Human CLDN7 aa 162-211/211. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 6-G3 | |
| Unconjugated | |
| CLDN7 | |
| CEPTRL2; claudin 7; claudin-1; Claudin-7; Cldn7; CLDN-7; clostridium perfringens enterotoxin receptor-like 2; CPETRL2; Hs.84359 | |
| Mouse | |
| Protein A | |
| RUO | |
| 1366, 53624, 65132 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction