Learn More
Invitrogen™ Claudin 7 Monoclonal Antibody (5D10F3)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ 374800
Product Code. 10537403
  • £403.00 / 100µg
For Research Use Only. All usage must comply with product instructions.
Description
Description
Claudin 7 Monoclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA, IP
This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions. Differential expression of this gene has been observed in different types of malignancies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claudin 7
Monoclonal
0.5 mg/mL
PBS with 0.1% sodium azide; pH 7.4
O95471, Q9Z261
CLDN7
Synthetic peptide derived from the C-terminal region of the human claudin-7 protein.
100 μg
Primary
Human, Mouse
Antibody
IgG2a κ
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
5D10F3
Unconjugated
CLDN7
CEPTRL2; claudin 7; claudin-1; Claudin-7; Cldn7; CLDN-7; clostridium perfringens enterotoxin receptor-like 2; CPETRL2; Hs.84359
Mouse
Protein A
RUO
1366, 53624
-20°C