missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Description
Claudin-6 Polyclonal antibody specifically detects Claudin-6 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Claudin-6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 50% glycerol, pH7.3 |
| Gene Alias | claudin 6, claudin-6, skullin |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 121-220 of human Claudin-6 (NP_067018.2). |
| Purification Method | Affinity purified |
| Quantity | 20 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?