missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 6 Monoclonal Antibody (8G5D2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538499
This item is not returnable.
View return policy
Description
This antibody has been tested in indirect ELISA.
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. These junctions are comprised of sets of continuous networking strands in the outwardly facing cytoplasmic leaflet, with complementary grooves in the inwardly facing extracytoplasmic leaflet. This gene encodes a component of tight junction strands, which is a member of the claudin family. The protein is an integral membrane protein and is one of the entry cofactors for hepatitis C virus. The gene methylation may be involved in esophageal tumorigenesis. This gene is adjacent to another family member CLDN9 on chromosome 16.
Specifications
| Claudin 6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P56747 | |
| CLDN6 | |
| Purified recombinant fragment of human CLDN6 (aa: extra(29-81) and extra(138-160)) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| 8G5D2 | |
| Unconjugated | |
| CLDN6 | |
| AL024037; claudin 6; Claudin6; claudin-6; CLD6; CLDN6; Skullin; tight junction molecule claudin 6; UNQ757/PRO1488 | |
| Mouse | |
| Protein G | |
| RUO | |
| 9074 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction