missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Claudin-5 Antibody (SR3444), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27772-100ul
This item is not returnable.
View return policy
Description
Claudin-5 Monoclonal antibody specifically detects Claudin-5 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| Claudin-5 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| SR3444 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| AWALTransmembrane protein deleted in VCFS, BEC1, claudin 5, claudin-5, CPETRL1, TMVCFTMDVCF, transmembrane protein deleted in velocardiofacial syndrome | |
| A synthesized peptide derived from human Claudin-5 (Uniprot #: O00501) | |
| 100 μL | |
| Cellular Markers | |
| 7122 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction