missing translation for 'onlineSavingsMsg'
Learn More

Claudin-5 Antibody (OTI1G4), DyLight 594, Novus Biologicals™

Product Code. 18406397 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406397 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406397 Supplier Novus Biologicals Supplier No. NBP271329DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Claudin-5 Monoclonal antibody specifically detects Claudin-5 in Human, Mouse, Rat, Porcine samples. It is validated for Flow Cytometry, Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin-5
Applications Flow Cytometry, Radioimmune Assays (RIA)
Classification Monoclonal
Clone OTI1G4
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias AWALTransmembrane protein deleted in VCFS, BEC1, claudin 5, claudin-5, CPETRL1, TMVCFTMDVCF, transmembrane protein deleted in velocardiofacial syndrome
Host Species Mouse
Immunogen Full length recombinant protein of human Claudin-5 (NP_003268) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7122
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.