missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 3 Monoclonal Antibody (KAA346_13F9), Invitrogen™
Mouse Monoclonal Antibody
£453.00 - £760.00
Specifications
| Antigen | Claudin 3 |
|---|---|
| Clone | KAA346_13F9 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30208616
|
Invitrogen™
MA555860 |
50 μL |
£453.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30194931
|
Invitrogen™
MA556240 |
200 μL |
£760.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Claudin 3 Monoclonal antibody specifically detects Claudin 3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| Claudin 3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AI182374; C7orf1; claudin 3; claudin-3; CLDN3; clostridium perfringens enterotoxin receptor 2; CPE-R 2; CPE-R2; CPE-receptor 2; CPETR2; hRVP1; integral membrane protein claudin-3; mRVP1; Rat ventral prostate.1 protein; Rat ventral prostate.1 protein homolog; RVP1; tight junction protein; ventral prostate.1 protein; ventral prostate.1 protein homolog; ventral prostate.1-like protein | |
| Human CLDN3 peptide (proprietary range information). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| CLDN3 |
| KAA346_13F9 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O15551 | |
| 1365 | |
| IgG2a | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title