Learn More
Claudin-3 Antibody (CL13128) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18577AF532
Product Code. 30290925
  • £582.00 / 0.10mL
Description
Claudin-3 Monoclonal antibody specifically detects Claudin-3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Claudin-3
Monoclonal
Alexa Fluor 532
C7orf1, claudin 3, claudin-3, Clostridium perfringens enterotoxin receptor 2, CPE-R 2, CPE-R2, CPETR2CPE-receptor 2, HRVP1, Rat ventral prostate.1 protein homolog, RVP1, ventral prostate.1 protein homolog, ventral prostate.1-like protein
This antibody was developed using a synthetic peptide derived from O15551, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cellular Markers, Extracellular Matrix
1365
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL13128
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified