missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Claudin 20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103746
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CLDN20.
The Claudin 20 gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions.
Specifications
| Claudin 20 | |
| Polyclonal | |
| Unconjugated | |
| CLDN20 | |
| claudin 20; claudin-20; CLDN20; EG621628; testis secretory sperm-binding protein Li 229n | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 49861, 680178 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P56880 | |
| CLDN20 | |
| A synthesized peptide derived from human CLDN20(Accession P56880), corresponding to amino acid residues I166-K216. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction