missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-11 Antibody (398205R), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4280RCL1
This item is not returnable.
View return policy
Description
Claudin-11 Monoclonal antibody specifically detects Claudin-11 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Claudin-11 | |
| Monoclonal | |
| CoraFluor 1 | |
| claudin 11, claudin-11, Oligodendrocyte-specific protein, OSPOTMoligodendrocyte transmembrane protein | |
| NS0 mouse myeloma cell line transfected with human Claudin-11, Met1-Val207, Accession # O75508 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 398205R | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5010 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction