missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92587-0.02ml
This item is not returnable.
View return policy
Description
Claudin 10 Polyclonal antibody specifically detects Claudin 10 in Rat samples. It is validated for Western Blot
Specifications
| Claudin 10 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| claudin 10, claudin-10, CPETRL3, OSP-L, OSP-like protein | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CLDN10 (NP_008915.1). MASTASEIIAFMVSISGWVLVSSTLPTDYWKVSTIDGTVITTATYWANLWKACVTDSTGVSNCKDFPSMLALDGYIQACRGLMIAAVSLGFFGSIFALFG | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 9071 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction