Learn More
Claudin-1 Rat anti-Human, Brilliant Violet 421, Clone: 421203, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 748199
This item is not returnable.
View return policy
Description
Claudin-1 Monoclonal antibody specifically detects Claudin-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Claudin-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| Claudin-1 | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 421203 | |
| Brilliant Violet 421 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG2a κ |