Learn More
Claudin 1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28674-1-AP-20UL
Description
The Claudin 1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Claudin 1. This antibody recognizes human, mouse, rat antigen. The Claudin 1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Claudin 1 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 9076 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |