missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516833
This item is not returnable.
View return policy
Description
PA5-16833 targets Claudin 1 in IHC (P) application and shows reactivity with Human, mouse, and Rat samples. The PA5-16833 immunogen is a synthetic peptide derived from the C-terminus of human Claudin-1.
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.
Specifications
| Claudin 1 | |
| Polyclonal | |
| PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
| O95832, O88551, P56745 | |
| CLDN1 | |
| A synthetic peptide derived from the C-terminus of human Claudin-1 | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat, Non-human Primate | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| CLDN1 | |
| claudin-1, senescence-associated epithelial membrane protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12737, 9076, 65129 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction