missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clathrin Light Chain A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584037
This item is not returnable.
View return policy
Description
Immunogen sequence: NDEAFAILDG GAPGPQPHGE PPGGPDAVDG VMNGEYYQES NGPTDSYAAI SQVDRLQSEP E Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 97%.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
Specifications
| Clathrin Light Chain A | |
| Polyclonal | |
| Unconjugated | |
| CLTA | |
| AV026556; Clathrin light chain A; clathrin, light chain (Lca); clathrin, light chain A; clathrin, light polypeptide (Lca); clathryn light chain (LCA2); Clta; Lca; LCA1; LCA2; LCA3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1211 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P09496 | |
| CLTA | |
| Recombinant protein corresponding to Human CLTA. Recombinant protein control fragment (Product #RP-103086). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction