missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clathrin light chain Antibody (SPM174), Janelia Fluor™ 646, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Clathrin light chain Monoclonal antibody specifically detects Clathrin light chain in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
Specifications
| Antigen | Clathrin light chain |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SPM174 |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca) |
| Host Species | Mouse |
| Immunogen | Purified recombinant N-terminal fragment of human Clathrin Light Chain |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?