missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clathrin light chain Antibody (CLC/1421), Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £409.00
Specifications
| Antigen | Clathrin light chain |
|---|---|
| Clone | CLC/1421 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18197989
|
Novus Biologicals
NBP2-53231-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18200970
|
Novus Biologicals
NBP2-53231-100UG |
100 μg |
£409.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Clathrin light chain Monoclonal specifically detects Clathrin light chain in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Clathrin light chain | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| Unconjugated | |
| Mouse | |
| Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 1211 | |
| IgG2b κ | |
| Protein A or G purified | |
| 37 kDa |
| CLC/1421 | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca) | |
| Purified recombinant N-terminal fragment of human Clathrin Light Chain | |
| Primary | |
| Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title