missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clathrin light chain Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92434-0.1ml
This item is not returnable.
View return policy
Description
Clathrin light chain Polyclonal antibody specifically detects Clathrin light chain in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Clathrin light chain | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-218 of human CLTA (NP_001824.1). MAELDPFGAPAGAPGGPALGNGVAGAGEEDPAAAFLAQQESEIAGIENDEAFAILDGGAPGPQPHGEPPGGPDAVDGVMNGEYYQESNGPTDSYAAISQVDRLQSEPESIRKWREEQMERLEALDANSRKQEAEWKEKAIKELEEWYARQDEQLQKTKANNRAAEEAFVNDIDESSPGTEWERVARLCDFNPKSSKQAKDVSRMRSVLISLKQAPLVH | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
| 1211 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction