missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Clathrin Heavy Chain Recombinant Rabbit Monoclonal Antibody (K01_2L86)
GREENER_CHOICE

Product Code. 30830106
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830106 100 μL
30825783 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830106 Supplier Invitrogen™ Supplier No. MA564024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: GRKQLLEKWL KEDKL.

Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clathrin Heavy Chain
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2L86
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene CLTC
Gene Accession No. P53675, Q00610
Gene Alias 3110065L21Rik; CHC; CHC17; CHC22; clathrin heavy chain; clathrin heavy chain 1; Clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 17; Clathrin heavy chain on chromosome 22; clathrin, heavy chain (Hc); clathrin, heavy chain-like 1; clathrin, heavy polypeptide (Hc); Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; clathrin, heavy polypeptide-like 2; CLH17; CLH-17; CLH22; CLH-22; Cltc; CLTCL; CLTCL1; CLTCL2; CLTD; Hc; KIAA0034; R74732
Gene Symbols CLTC, CLTCL1
Host Species Rabbit
Immunogen A synthetic peptide of human Clathrin heavy chain (aa 443-457).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1213, 8218
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.