missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clathrin Heavy Chain Recombinant Rabbit Monoclonal Antibody (23GB3475), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Clathrin Heavy Chain |
|---|---|
| Clone | 23GB3475 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088035
![]() |
Invitrogen™
MA553622 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089287
|
Invitrogen™
MA553623 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Clathrin Heavy Chain Recombinant Monoclonal antibody specifically detects Clathrin Heavy Chain in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Clathrin Heavy Chain | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P11442, P53675, Q00610, Q68FD5 | |
| 1213, 54241, 67300, 8218 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3475 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 3110065L21Rik; CHC; CHC17; CHC22; clathrin heavy chain; clathrin heavy chain 1; Clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 17; Clathrin heavy chain on chromosome 22; clathrin, heavy chain (Hc); clathrin, heavy chain-like 1; clathrin, heavy polypeptide (Hc); Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; clathrin, heavy polypeptide-like 2; CLH17; CLH-17; CLH22; CLH-22; Cltc; CLTCL; CLTCL1; CLTCL2; CLTD; Hc; KIAA0034; R74732 | |
| A synthetic peptide of human Clathrin heavy chain (204-256AA). | |
| Primary | |
| -20°C | |
| CLTCL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title