missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Clathrin Heavy Chain Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584404
This item is not returnable.
View return policy
Description
Immunogen sequence: ESLRKEEEQA TETQPIVYGQ PQLMLTAGPS VAVPPQ Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
Specifications
| Clathrin Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| CLTCL1 | |
| 3110065L21Rik; CHC; CHC17; CHC22; clathrin heavy chain; clathrin heavy chain 1; Clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 17; Clathrin heavy chain on chromosome 22; clathrin, heavy chain (Hc); clathrin, heavy chain-like 1; clathrin, heavy polypeptide (Hc); Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; clathrin, heavy polypeptide-like 2; CLH17; CLH-17; CLH22; CLH-22; Cltc; CLTCL; CLTCL1; CLTCL2; CLTD; Hc; KIAA0034; R74732 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1213, 8218 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53675, Q00610 | |
| CLTC, CLTCL1 | |
| Recombinant protein corresponding to Human CLTC. Recombinant protein control fragment (Product #RP-103974). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido