missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Clathrin Heavy Chain Antibody (Academia Sinica patent anti-Clathrin Heavy Chain) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28577-1mg
This item is not returnable.
View return policy
Description
Clathrin Heavy Chain Monoclonal antibody specifically detects Clathrin Heavy Chain in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Clathrin Heavy Chain | |
| Monoclonal | |
| Unconjugated | |
| Q00610 | |
| Clathrin Heavy Chain/ CHC | |
| 1 mg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Academia Sinica patent anti-Clathrin Heavy Chain | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Human | |
| Protein A purified | |
| RUO | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction