missing translation for 'onlineSavingsMsg'
Learn More

Clathrin Heavy Chain 2/CHC22/CLTCL1 Antibody [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30736546 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736546 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736546 Supplier Novus Biologicals Supplier No. AF6948J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Clathrin Heavy Chain 2/CHC22/CLTCL1 Polyclonal antibody specifically detects Clathrin Heavy Chain 2/CHC22/CLTCL1 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clathrin Heavy Chain 2/CHC22/CLTCL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias CHC22, clathrin heavy chain 2, clathrin heavy chain on chromosome 22, clathrin, heavy chain-like 1, Clathrin, heavy polypeptide D, CLTCL, heavy polypeptide-like 1
Host Species Sheep
Immunogen E. coli-derived recombinant human Clathrin Heavy Chain 2/CHC22, Trp1521-Glu1640, Accession # P53675
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8218
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.